Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

792675

Sigma-Aldrich

2-Fluoro-5-chlorobenzenesulfonyl fluoride

97%

Synonym(s):

5-Chloro-2-fluorobenzenesulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3ClF2O2S
CAS Number:
Molecular Weight:
212.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

mp

35-41 °C

functional group

chloro
fluoro

SMILES string

FC1=C(S(F)(=O)=O)C=C(Cl)C=C1

InChI

1S/C6H3ClF2O2S/c7-4-1-2-5(8)6(3-4)12(9,10)11/h1-3H

InChI key

DOVCXFSROGOKQI-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service