Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79231

Sigma-Aldrich

(R)-(−)-α-(Trifluoromethyl)benzyl alcohol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym(s):

(−)-Phenyl(trifluoromethyl)carbinol, (R)-(−)-1-Phenyl-2,2,2-trifluoroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CF3)OH
CAS Number:
Molecular Weight:
176.14
Beilstein:
2413959
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D −31.5±0.5°, neat

optical purity

enantiomeric ratio: ≥98:2 (GC)

refractive index

n20/D 1.459 (lit.)
n20/D 1.462

bp

73-76 °C/9 mmHg (lit.)

mp

20 °C (lit.)

density

1.300 g/mL at 20 °C (lit.)

SMILES string

O[C@H](c1ccccc1)C(F)(F)F

InChI

1S/C8H7F3O/c9-8(10,11)7(12)6-4-2-1-3-5-6/h1-5,7,12H/t7-/m1/s1

InChI key

VNOMEAQPOMDWSR-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral solvent for the determination of enantiomeric composition by NMR ; Chiral auxiliary

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Bucciarelli et al.
Journal of the Chemical Society. Perkin Transactions 1, 2152-2152 (1980)
A. Forni et al.
Journal of the Chemical Society. Chemical Communications, 588-588 (1981)
Pirkle, W.H. and Rinaldi, P.L.
The Journal of Organic Chemistry, 42, 3217-3217 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service