Skip to Content
MilliporeSigma
All Photos(2)

Documents

792047

Sigma-Aldrich

2-Bromoisobutanoic acid N-hydroxysuccinimide ester

98% (GC)

Synonym(s):

1-(2-Bromo-2-methyl-1-oxopropoxy)-2,5-pyrrolidinedione, 2,5-Dioxopyrrolidin-1-yl 2-bromo-2-methylpropanoate, 2-Bromo-2-methyl-propanoic acid 2,5-dioxo-1-pyrrolidinyl ester, 2-Bromoisobutyryl(N-hydroxysuccinimide), N-Hydroxysuccinimidyl 2-bromo-2-methylpropionate, NHS ATRP initiator, NHS protected Bromoisobutyrate, NHS-BiB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BrNO4
CAS Number:
Molecular Weight:
264.07
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98% (GC)

form

liquid

mp

135-140 °C

storage temp.

2-8°C

SMILES string

O=C1N(OC(C(C)(C)Br)=O)C(CC1)=O

InChI

1S/C8H10BrNO4/c1-8(2,9)7(13)14-10-5(11)3-4-6(10)12/h3-4H2,1-2H3

InChI key

QBJWGGWPQPRVKW-UHFFFAOYSA-N

Related Categories

Application

Atom transfer radical polymerization (ATRP) initiator with an NHS ester moiety for conjugation chemistry, useful for biological ligations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bailey Risteen et al.
Small (Weinheim an der Bergstrasse, Germany), 14(46), e1802060-e1802060 (2018-09-11)
A thermally "switchable" liquid-crystalline (LC) phase is observed in aqueous suspensions of cellulose nanocrystals (CNCs) featuring patchy grafts of the thermoresponsive polymer poly(N-isopropylacrylamide) (PNIPAM). "Patchy" polymer decoration of the CNCs is achieved by preferential attachment of an atom transfer radical
Bhalchandra S Lele et al.
Biomacromolecules, 6(6), 3380-3387 (2005-11-15)
We have developed a novel technique to synthesize near-uniform protein-polymer conjugates by initiating atom transfer radical polymerization of monomethoxy poly(ethylene glycol)-methacrylate from 2-bromoisobutyramide derivatives of chymotrypsin (a protein-initiator). Polymerization initiated from the monosubstituted protein-initiator resulted in the conjugate containing a
Shuaidong Huo et al.
Nature chemistry, 13(2), 131-139 (2021-01-31)
Pharmaceutical drug therapy is often hindered by issues caused by poor drug selectivity, including unwanted side effects and drug resistance. Spatial and temporal control over drug activation in response to stimuli is a promising strategy to attenuate and circumvent these

Articles

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service