Skip to Content
MilliporeSigma
All Photos(1)

Documents

791466

Sigma-Aldrich

EL-HSE high stability electrolyte

Synonym(s):

EL-HSE, EL-HSE electrolyte, Greatcell Solar®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

form

liquid

color

semi-transparent amber

refractive index

n20/D 1.442

conductivity

8-10 mS/cm at 20 °C

density

1.034 g/mL at 25 °C

Application

EL-HSE High Stability Electrolyte is a low toxicity liquid electrolyte to achieve ultra-long life in normal operating conditions.
This is the “work horse” electrolyte system for long term stability testing. Particularly suitable for glass or metal/glass substrate-based DSC systems.

Outstanding stability has been achieved with EL-HSE, 25,600 hours of continuous light soaking at 55-60°C and 3,000 hours at 85°C.

Legal Information

Product of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar Materials Pty Ltd.
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

151.0 °F

Flash Point(C)

66.11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of large area photovoltaic dye cells at 3GSolar
Jonathan Goldstein; Ilya Yakupov; Barry Breen;
Solar Energy Materials and Solar Cells, 94, 638-641 (2010)

Articles

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service