Skip to Content
MilliporeSigma
All Photos(1)

Documents

791261

Sigma-Aldrich

L-Proline-2,5,5-d3

98 atom % D, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D3H6NO2
CAS Number:
Molecular Weight:
118.15
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97%

form

solid

technique(s)

bio NMR: suitable

mass shift

M+3

SMILES string

[2H]C1([2H])CC[C@]([2H])(C(O)=O)N1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m1/s1/i3D2,4D

InChI key

ONIBWKKTOPOVIA-LTDLRDEHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rika Shiota et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 33(6), 719-722 (2017-06-13)
An analytical method was developed and validated for the measurement of hydroxyproline (Hyp) levels in mouse kidney by high-performance liquid chromatography with tandem mass spectrometric detection (LC/MS/MS) using an analytical column specially designed for the LC/MS/MS analysis for intact amino
Holly Newton et al.
Nature communications, 11(1), 4653-4653 (2020-09-18)
Cancer cells demand excess nutrients to support their proliferation, but how tumours exploit extracellular amino acids during systemic metabolic perturbations remain incompletely understood. Here, we use a Drosophila model of high-sugar diet (HSD)-enhanced tumourigenesis to uncover a systemic host-tumour metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service