Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

790796

Sigma-Aldrich

Zinc benzylsulfinate

Synonym(s):

Zinc benzylsulfinate, Baran BNS Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O4S2Zn
CAS Number:
Molecular Weight:
375.78
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Alkylations
reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

reagent type: diversification reagent

mp

56 °C (Decomposition)

storage temp.

2-8°C

SMILES string

O=S(O[Zn]OS(CC1=CC=CC=C1)=O)CC2=CC=CC=C2

InChI

1S/2C7H8O2S.Zn/c2*8-10(9)6-7-4-2-1-3-5-7;/h2*1-5H,6H2,(H,8,9);/q;;+2/p-2

InChI key

AHSIXHPQMGLVIG-UHFFFAOYSA-L

Application

Zinc benzylsulfinate (BNS) is part of a zinc sulfinate toolbox developed by Phil Baran for the simple and rapid diversification of heterocycles.


Practical and Innate Carbon-Hydrogen Functionalization of Heterocycles

Learn More at the Professor and Product Portal of Professor Phil S. Baran.

Other Notes

may contain up to 1 mole eq ZnCl2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fionn O'Hara et al.
Journal of the American Chemical Society, 135(32), 12122-12134 (2013-07-19)
Radical addition processes can be ideally suited for the direct functionalization of heteroaromatic bases, yet these processes are only sparsely used due to the perception of poor or unreliable control of regiochemistry. A systematic investigation of factors affecting the regiochemistry
Yuta Fujiwara et al.
Nature, 492(7427), 95-99 (2012-12-04)
Nitrogen-rich heterocyclic compounds have had a profound effect on human health because these chemical motifs are found in a large number of drugs used to combat a broad range of diseases and pathophysiological conditions. Advances in transition-metal-mediated cross-coupling have simplified

Articles

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service