Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

790400

Sigma-Aldrich

Potassium 2-chloro-4-(trifluoromethyl)pyridine-3-trifluoroborate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2BClF6KN
Molecular Weight:
287.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

330-333 °C

SMILES string

ClC1=C([B-](F)(F)F)C(C(F)(F)F)=CC=N1.[K+]

InChI

1S/C6H2BClF6N.K/c8-5-4(7(12,13)14)3(1-2-15-5)6(9,10)11;/h1-2H;/q-1;+1

InChI key

LQQDLQPZJJRMON-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service