Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78972

Sigma-Aldrich

(R)-1-Phenyl-2-propen-1-ol

technical, ≥95% (sum of enantiomers, GC)

Synonym(s):

(R)-α-Vinylbenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O
CAS Number:
Molecular Weight:
134.18
Beilstein:
4658061
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

technical

Assay

≥95% (sum of enantiomers, GC)

optical activity

[α]20/D +1.0±0.5°, c = 3.2% in chloroform

optical purity

enantiomeric ratio: ≥93:7 (GC)

refractive index

n20/D 1.543

density

1.021 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

O[C@H](C=C)c1ccccc1

InChI

1S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h2-7,9-10H,1H2/t9-/m1/s1

InChI key

MHHJQVRGRPHIMR-SECBINFHSA-N

Other Notes

Chiral building block

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.A. Claremon et al.
Journal of the American Chemical Society, 108, 8265-8265 (1986)
Lin Yi et al.
Analytical and bioanalytical chemistry, 386(3), 666-674 (2006-05-26)
Characterizing the biological effects of metabolic transformations (or biotransformation) is one of the key steps in developing safe and effective pharmaceuticals. Sulfate conjugation, one of the major phase II biotransformations, is the focus of this study. While this biotransformation typically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service