Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78172

Sigma-Aldrich

(R)-(−)-Phenylsuccinic acid

≥96.0% (sum of enantiomers, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
Beilstein:
3201702
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (sum of enantiomers, HPLC)

form

powder

optical activity

[α]20/D −175±4°, c = 1% in acetone

mp

173-176 °C

SMILES string

OC(=O)C[C@@H](C(O)=O)c1ccccc1

InChI

1S/C10H10O4/c11-9(12)6-8(10(13)14)7-4-2-1-3-5-7/h1-5,8H,6H2,(H,11,12)(H,13,14)/t8-/m1/s1

InChI key

LVFFZQQWIZURIO-MRVPVSSYSA-N

Other Notes

Chiral building block; preparation of 3-phenylpyrrolidine; reduction to 2-phenyl-1,4-butanediol and synthesis of the dibromide; preparation of 3-oxo-1-indanecarboxylic acid

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G. Bettoni et al.
Journal of Heterocyclic Chemistry, 13, 1053-1053 (1976)
H.-J. Hansen et al.
Helvetica Chimica Acta, 65, 325-325 (1982)
H.-J. Hansen et al.
Helvetica Chimica Acta, 62, 1120-1120 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service