Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779741

Sigma-Aldrich

Methyl 2-azidobenzoate solution

~0.5 M in tert-butyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
Beilstein:
475220
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

COC(=O)c1ccccc1N=[N+]=[N-]

InChI

1S/C8H7N3O2/c1-13-8(12)6-4-2-3-5-7(6)10-11-9/h2-5H,1H3

InChI key

LGHLODHMWICPLN-UHFFFAOYSA-N

Application

Review

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-27.4 °F

Flash Point(C)

-33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Bräse et al.
Angewandte Chemie (International ed. in English), 44(33), 5188-5240 (2005-08-16)
Since the discovery of organic azides by Peter Griess more than 140 years ago, numerous syntheses of these energy-rich molecules have been developed. In more recent times in particular, completely new perspectives have been developed for their use in peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service