Skip to Content
MilliporeSigma
All Photos(1)

Documents

779709

Sigma-Aldrich

1-Azido-4-(trifluoromethoxy) benzene solution

0.25 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4F3N3O
CAS Number:
Molecular Weight:
203.12
Beilstein:
10425646
MDL number:
UNSPSC Code:
12352200

Assay

≥95.0% (HPLC)

concentration

0.25 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

FC(F)(F)Oc1ccc(cc1)N=[N+]=[N-]

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-27.4 °F

Flash Point(C)

-33 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingyu Hu et al.
Organic letters, 10(24), 5529-5531 (2008-12-05)
A panel of small molecule-based MMP inhibitors containing rhodanine warheads was assembled using "one-pot" click chemistry. Upon biological screening, moderate inhibitors were identified which specifically targets MMP-7 and MMP-13 over other MMPs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service