Skip to Content
MilliporeSigma
All Photos(2)

Documents

779636

Sigma-Aldrich

1-Bromo-2-naphthaldehyde

≥96.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7BrO
CAS Number:
Molecular Weight:
235.08
Beilstein:
2613639
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (HPLC)
96.0-104.0% (T)

form

crystals

SMILES string

Brc1c(C=O)ccc2ccccc12

InChI

1S/C11H7BrO/c12-11-9(7-13)6-5-8-3-1-2-4-10(8)11/h1-7H

InChI key

CYGUXEZVBLMVRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service