Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779512

Sigma-Aldrich

Pentafluorophenyl acetate

≥97.0% (GC)

Synonym(s):

Acetic acid pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3F5O2
CAS Number:
Molecular Weight:
226.10
Beilstein:
1985849
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

crystals

SMILES string

CC(=O)Oc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C8H3F5O2/c1-2(14)15-8-6(12)4(10)3(9)5(11)7(8)13/h1H3

InChI key

ZXTVBLZVILLKPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service