Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779407

Sigma-Aldrich

4-Pentyn-1-amine

≥92.0%

Synonym(s):

1-Amino-4-pentyne, 4-Pentynylamine, 5-Amino-1-pentyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2C≡CH
CAS Number:
Molecular Weight:
83.13
Beilstein:
2232239
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥92.0%

form

liquid

reaction suitability

reaction type: click chemistry

density

0.859 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

NCCCC#C

InChI

1S/C5H9N/c1-2-3-4-5-6/h1H,3-6H2

InChI key

LMDDPGHBJXJGAC-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 - 59.0 °F

Flash Point(C)

10 - 15 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolò Mauro et al.
International journal of pharmaceutics, 555, 207-219 (2018-11-21)
The extremely complex tumor microenvironment (TME) in humans is the major responsible for the therapeutic failure in cancer nanomedicine. A new concept of disease-driven nanomedicine, henceforth named "Theranomics", which attempts to target cancer cells and TME on the whole, represents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service