Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

778842

Sigma-Aldrich

1-Azido-2-fluorobenzene solution

0.5 M in tert-butyl methyl ether, ≥95% (HPLC)

Synonym(s):

o-Fluorophenyl azide solution, 2-Fluorophenyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4FN3
CAS Number:
Molecular Weight:
137.11
Beilstein:
2501863
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 778842, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

≥95% (HPLC)

concentration

0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

Fc1ccccc1N=[N+]=[N-]

InChI

1S/C6H4FN3/c7-5-3-1-2-4-6(5)9-10-8/h1-4H

InChI key

YOBOIKMFTLTNTR-UHFFFAOYSA-N

General description

1-Azido-2-fluorobenzene is an aromatic azide generally used in copper(I)-catalyzed azide-alkyne cycloaddition reactions.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F

Flash Point(C)

-28 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cu-catalyzed azide? alkyne cycloaddition
Meldal M & Torn?e CW
Chemical Reviews, 108(8), 2952-3015 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service