Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

777781

Sigma-Aldrich

7-Bromo-4-fluoro-1-indanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6BrFO
CAS Number:
Molecular Weight:
229.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

135-137 °C

functional group

bromo
fluoro
ketone

SMILES string

Fc1ccc(Br)c2C(=O)CCc12

InChI

1S/C9H6BrFO/c10-6-2-3-7(11)5-1-4-8(12)9(5)6/h2-3H,1,4H2

InChI key

AIVSTGKLEISXKC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service