Skip to Content
MilliporeSigma
All Photos(3)

Documents

777692

Sigma-Aldrich

(1,10-Phenanthroline)(trifluoromethyl)copper(I)

90%

Synonym(s):

Trifluoromethylator®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8CuF3N2
CAS Number:
Molecular Weight:
312.76
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

storage temp.

2-8°C

InChI

1S/C12H8N2.CF3.Cu/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;2-1(3)4;/h1-8H;;

InChI key

CUQZQRIIFNWQSJ-UHFFFAOYSA-N

General description

(1,10-Phenanthroline)(trifluoromethyl)copper(I) [(Phen)Cu-CF3] can be prepared by treating [CuOt-Bu]4 with 1,10-phenanthroline, followed by the addition of (trifluoromethyl)trimethylsilane (CF3TMS). It is an easily handled, thermally stable, single-component reagent.

Application

Trifluoromethylator, A New Arene Trifluoromethylation Reagent: (Phen)Cu-CF3

(Phen)Cu-CF3 can be used as a reagent for the trifluoromethylation of aryl iodides. Electron-rich and electron-deficient iodoarenes, as well as sterically-hindered iodoarenes, react in high yield under mild conditions. Electrophilic functional groups, including aldehydes, nitroarenes, ketones, and esters are tolerated. The substrate scope of this reagent is broad and the reagent has wide applicability for trifluoromethylation reactions.
It can also be used in the oxidative trifluoromethylation of organoboron reagents, and terminal alkynes through C-H activation.

Legal Information

Trifluoromethylator is a registered trademark of Catylix, Inc

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(1, 10-Phenanthroline)(trifluoromethyl) copper (I)
Qing F-L
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-4 (2001)
A broadly applicable copper reagent for trifluoromethylations and perfluoroalkylations of aryl iodides and bromides.
Hiroyuki Morimoto et al.
Angewandte Chemie (International ed. in English), 50(16), 3793-3798 (2011-03-29)

Related Content

Professor Hartwig's research focuses on the discovery and understanding of new reactions catalyzed by transition metal complexes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service