Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775924

Sigma-Aldrich

4-Benzoylbenzoic acid N-succinimidyl ester

97%

Synonym(s):

4-Benzoylbenzoic acid, succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13NO5
CAS Number:
Molecular Weight:
323.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

form

solid

mol wt

323.3

reaction suitability

reagent type: cross-linking reagent

mp

207-212 °C

storage temp.

−20°C

SMILES string

O=C1CCC(=O)N1OC(=O)c2ccc(cc2)C(=O)c3ccccc3

InChI

1S/C18H13NO5/c20-15-10-11-16(21)19(15)24-18(23)14-8-6-13(7-9-14)17(22)12-4-2-1-3-5-12/h1-9H,10-11H2

InChI key

MVQNJLJLEGZFGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A benzophenone-based photoreactive reagent shown to be useful for synthesizing photoreactive peptides and oligonucleotides.
Heterobifunctional photoreactive crosslinker. Succinimidyl ester will react with amines and the benzophenone will react non-specifically upon photoirradiation at approximately 360 nm.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dan Bernardi et al.
Bioorganic & medicinal chemistry letters, 16(6), 1601-1604 (2006-01-03)
The first synthesis of an N-acylated photoactivatable analogue of reduced glutathione is described. N-(4-Benzoylbenzoyl)glutathione (8) was found to be an inhibitor and a photoaffinity probe of purified rat liver glutathione S-transferases.
Horacio F Olivo et al.
Bioorganic & medicinal chemistry letters, 20(3), 1153-1155 (2009-12-25)
We have synthesized an analog of dehydroepiandrosterone (DHEA, 1) containing both a benzophenone (BP) and a biotin (Bt) group (DHEA-BP-Bt, 8). Compound 8 was prepared by functionalization on C-17 of 1. Biocytin was reacted with 4-benzoylbenzoic acid and the product

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service