Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

774847

Sigma-Aldrich

1,2,3,4-Tetrahydro-4-methylquinoline-8-carboxylic acid

97%

Synonym(s):

4-Methyl-1,2,3,4-tetrahydroquinoline-8-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO2
CAS Number:
Molecular Weight:
191.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

165-168 °C

functional group

carboxylic acid

SMILES string

CC1CCNc2c1cccc2C(O)=O

InChI

1S/C11H13NO2/c1-7-5-6-12-10-8(7)3-2-4-9(10)11(13)14/h2-4,7,12H,5-6H2,1H3,(H,13,14)

InChI key

YPHSTQBSRIRNQJ-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service