Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773468

Sigma-Aldrich

Propargyl 2-bromoisobutyrate

>97%

Synonym(s):

Acetylene ATRP initiator, Acetylene BiB, Propyne 2-bromoisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9BrO2
CAS Number:
Molecular Weight:
205.05
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

>97%

form

solid or liquid

bp

188-194 °C

SMILES string

CC(C)(Br)C(=O)OCC#C

InChI

1S/C7H9BrO2/c1-4-5-10-6(9)7(2,3)8/h1H,5H2,2-3H3

InChI key

XTILOBFYTKJHJZ-UHFFFAOYSA-N

Application

Atom transfer radical polymerization (ATRP) initiator with an acetylene functionality that enables Cu-mediated ligation (“click” chemistry) or other functionalization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.9 °F

Flash Point(C)

89.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haobo Sun et al.
Biosensors & bioelectronics, 131, 193-199 (2019-03-06)
Here we report a highly selective and ultrasensitive DNA biosensor based on electrochemical atom transfer radical polymerization (ATRP) signal amplification and "Click Chemistry". The DNA biosensor was prepared by immobilizing thiol and azide modified hairpin DNAs on gold electrode surface.
Xin Liu et al.
Polymers, 11(3) (2019-04-10)
Herein, we report the formation and characterization of novel amphiphilic linear-dendritic block copolymers (LDBCs) composed of hydrophilic dendritic poly(ether-ester), PEE, blocks and hydrophobic linear poly(styrene), PSt. The LDBCs are synthesized via controlled atom transfer radical polymerization (ATRP) initiated by a
Xiaoke Zheng et al.
Talanta, 207, 120290-120290 (2019-10-09)
Human T-lymphotropic virus type II (HTLV-II) is a crucial retrovirus that is closely associated with a variety of human diseases. Herein, an ultrasensitive fluorescent HTLV-II DNA detection strategy was developed for the first time based on magnetic nanoparticles (MNPs) and

Articles

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service