Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

773417

Sigma-Aldrich

Poly(vinyl pyrrolidone), cyanomethyl diphenylcarbamodithioate

average Mn 6,000, PDI ≤1.2

Synonym(s):

PVP, PVP-RAFT, Polyvidone, Povidone, poly(vinyl pyrrolidinone)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H2N(C6H9NO)nC13H10NS2
UNSPSC Code:
12352100
NACRES:
NA.23

form

powder

mol wt

average Mn 6,000

transition temp

Tg 146-150 °C

PDI

≤1.2

Application

Poly(vinyl pyrrolidone) is commonly used in biomedical applications. The end group functionality contains a cyanomethyl group; and the RAFT agent diphenylcarbamodithioate which will allow for additional polymerization to generate a diblock copolymer. This RAFT agent is well-suited for vinyl acetates and vinyl benzoate monomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service