Skip to Content
MilliporeSigma
All Photos(2)

Documents

773379

Sigma-Aldrich

Poly(2-ethyl-2-oxazoline) α-methyl, ω-2-hydroxyethylamine terminated

average Mn 5,000, PDI <1.3

Synonym(s):

POZ, Poly(2-ethyl-2-oxazoline) hydroxyethyl amine, amino terminated polyoxazoline, aminoPEOx, polyoxazoline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C5H9NO)nNHC2H4OH
UNSPSC Code:
12162002
NACRES:
NA.23

description

Tg 54°C

form

solid

mol wt

average Mn 5,000

PDI

<1.3

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Polyoxazolines with amine functionality can be covalently bound to carboxyl groups or sulfhydryl groups. Amines can also attach to oxidized carbohydrate groups (usually aldehydes) by reductive amination. Polyoxazolines are a biocompatible, hydrophilic, amorphous alternatives to poly(ethylene glycol).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Poly(2-oxazoline)s, synthesized via LCROP, resemble polypeptides and have various applications in polymer chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service