Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772534

Sigma-Aldrich

Ethyl 2-(trifluoromethyl)thiazole-5-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6F3NO2S
CAS Number:
Molecular Weight:
225.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.441

density

1.344 g/mL at 25 °C

functional group

ester
fluoro

SMILES string

CCOC(=O)c1cnc(s1)C(F)(F)F

InChI

1S/C7H6F3NO2S/c1-2-13-5(12)4-3-11-6(14-4)7(8,9)10/h3H,2H2,1H3

InChI key

KQTWGECEHUVSPX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

198.0 °F

Flash Point(C)

92.22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service