Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

772496

Sigma-Aldrich

3-Bromo-2-(difluoromethyl)pyridine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrF2N
CAS Number:
Molecular Weight:
208.00
UNSPSC Code:
12352100

Assay

95%

form

liquid

refractive index

n20/D 1.522

density

1.717 g/mL at 25 °C

InChI

1S/C6H4BrF2N/c7-4-2-1-3-10-5(4)6(8)9/h1-3,6H

InChI key

MDRUEFNPMGSINM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service