Skip to Content
MilliporeSigma
All Photos(1)

Documents

771732

Sigma-Aldrich

2-Nitrophenol-13C6

99 atom % 13C, 99% (CP)

Synonym(s):

o-Nitrophenol-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6H5NO3
Molecular Weight:
145.06
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

mass shift

M+6

SMILES string

O[13C]1=[13C]([N+]([O-])=O)[13CH]=[13CH][13CH]=[13CH]1

InChI

1S/C6H5NO3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

IQUPABOKLQSFBK-IDEBNGHGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service