Skip to Content
MilliporeSigma
All Photos(1)

Documents

771260

Sigma-Aldrich

3,4-Dimethoxyphenylboronic acid MIDA ester

95%

Synonym(s):

2-(3,4-Dimethoxyphenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16BNO6
Molecular Weight:
293.08
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

235-239 °C

SMILES string

O=C(CN(C)C1)OB(C2=CC(OC)=C(OC)C=C2)OC1=O

InChI

1S/C13H16BNO6/c1-15-7-12(16)20-14(21-13(17)8-15)9-4-5-10(18-2)11(6-9)19-3/h4-6H,7-8H2,1-3H3

InChI key

ZOFITLKPVZEWMX-UHFFFAOYSA-N

Application

Protected boronic acid with methylimindodiacetic acid (MIDA) that does not react under standard Suzuki-Miyuara coupling conditions, allowing for selective and iterative cross-coupling sequences. The MIDA boronate also provides prolonged shelf life and stability to typically unstable boronic acid derivatives.

MIDA Boronates

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service