769924
Resomer® L 210 S, Poly(L-lactide)
ester terminated
Synonym(s):
Resomer® L 210S, PLLA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
form
granular
viscosity
3.3-4.3 dL/g
mp
180-185 °C
transition temp
Tg 60-65 °C
storage temp.
2-8°C
InChI
1S/C6H8O4/c1-3-5(7)10-4(2)6(8)9-3/h3-4H,1-2H3/t3-,4-/m0/s1
InChI key
JJTUDXZGHPGLLC-IMJSIDKUSA-N
General description
Resomer® L 210 S, Poly(L-lactide) (PLLA), a high-performance bio-based polymer, can be prepared from corn starch. Its properties include biodegradability, hydrophobicity, and excellent composting characteristics.
Application
Biodegradable, biocompatible, and bioresorbable polymer composed of L-Lactide. This semi-crystalline material has been used in the fabrication of research medical devices and research tissue engineering solutions, such as orthopedic or soft tissue fixation devices. Degradation of this materials has been thoroughly studied and has been shown to be safely resorbed by the body after implantation. Modification of molecular weight and polymer composition allows for control of the degradation rate and mechanical stability of the polymer.
PLLA can be used as a resorbable implant material for potential usage in biomedical applications. It can also be used in the formation of composite ceramic material for biological applications.
Legal Information
RESOMER is a registered trademark of Evonik Rohm GmbH
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stereocomplexed polylactides (Neo-PLA) as high-performance bio-based polymers: their formation, properties, and application
Polymer International, 55(6), 626-642 (2006)
Poly (L-lactide)(PLLA)/multiwalled carbon nanotube (MWCNT) composite: characterization and biocompatibility evaluation
The Journal of Physical Chemistry B, 110(26), 12910-12915 (2006)
Synthesis and properties of hydroxyapatite/poly-L-lactide composite biomaterials
Biomaterials, 20(9), 809-816 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service