Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769258

Sigma-Aldrich

Zirconium(IV) oxide

granular, 3-6 mm

Synonym(s):

Zirconia

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrO2
CAS Number:
Molecular Weight:
123.22
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

form

granular

impurities

≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)

particle size

3-6 mm

bp

5000 °C (lit.)

mp

2700 °C (lit.)

density

5.89 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%
Co: ≤0.0005%
Cr: ≤0.002%
Cu: ≤0.001%
Fe: ≤0.002%
Hg: ≤0.1%
Pb: ≤0.1%
Ti: ≤0.2%
V: ≤0.005%

SMILES string

O=[Zr]=O

InChI

1S/2O.Zr

InChI key

MCMNRKCIXSYSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Zirconium Dioxide, ZrO2, is a high refractive index, low absorption material used for optical coatings in the near-UV to IR spectral regions.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A high-performance spectrally-selective solar absorber based on a yttria-stabilized zirconia cermet with high-temperature stability
Cao, F., et al.
Energy & Environmental Science, 8, 3040-3048 (2015)
Silica?zirconia alkali-resistant coatings by sol?gel route
Dhere, SL.
Current Science, 108(9), 1647-1652 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service