Skip to Content
MilliporeSigma
All Photos(2)

Documents

769029

Sigma-Aldrich

Neon

Messer® CANGas, 99.999%

Synonym(s):

Neon gas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ne
CAS Number:
Molecular Weight:
20.18
EC Number:
MDL number:
UNSPSC Code:
12142006
PubChem Substance ID:
NACRES:
NA.22

vapor density

0.7 (21 °C, vs air)

Quality Level

description

Filling mass: 10g
Filling volume: 12L

Assay

99.999%

form

gas

resistivity

6.97 μΩ-cm, 20°C

bp

−246 °C (lit.)

mp

-248.67 °C (lit.)

SMILES string

[Ne]

InChI

1S/Ne

InChI key

GKAOGPIIYCISHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Neon is a colourless and odourless rare gas that is lighter than air. Applications include use as a filling gas for tubular neon lamps or buoyancy gas for stratospheric balloons. Neon is also a component of laser gas mixtures (helium-neon lasers, excimer lasers) and panning gas (filling gas for plasma display panels [PDP]). Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Recommended products

Z742374: CANgas pressure regulator
Z742379: CANgas plastic hose spout
Z742378: CANgas outer adaptor
Z742377: CANgas inner adaptor
Z742375: CANgas dosing valve

Legal Information

Messer is a registered trademark of Messer Group GmbH

accessory

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service