Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

768855

Sigma-Aldrich

7-Bromo-3,4-dihydroisoquinoline

98.5-100% (GC)

Synonym(s):

7-Bromo-3,4-dihydroisoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8BrN
CAS Number:
Molecular Weight:
210.07
Beilstein:
1238319
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98.5-100% (GC)

form

solid

suitability

corresponds for identity (IR)

functional group

bromo

SMILES string

BrC1=CC=C2C(C=NCC2)=C1

InChI

1S/C9H8BrN/c10-9-2-1-7-3-4-11-6-8(7)5-9/h1-2,5-6H,3-4H2

InChI key

YQQLGLRTMLGKDQ-UHFFFAOYSA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service