Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

768553

Sigma-Aldrich

Bis[2-(2-benzothiazolyl-N3)phenolato-O]zinc

99%

Synonym(s):

Zn(BTZ)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16N2O2S2Zn
CAS Number:
Molecular Weight:
517.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

mp

305-310 °C

λmax

334+/-5 nm at 0.01 g/L in dichloromethane

SMILES string

O([Zn]Oc1ccccc1-c2nc3ccccc3s2)c4ccccc4-c5nc6ccccc6s5

InChI

1S/2C13H9NOS.Zn/c2*15-11-7-3-1-5-9(11)13-14-10-6-2-4-8-12(10)16-13;/h2*1-8,15H;/q;;+2/p-2

InChI key

CJGUQZGGEUNPFQ-UHFFFAOYSA-L

Application

  • Green Light Emitters and Dopants
  • Metal Complex Dopants and Dyes
  • OLED and PLED Materials
  • Organic and Printed Electronics
  • Photoluminescent Materials
  • Photonic and Optical Materials

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service