Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

768405

Sigma-Aldrich

9,9′-[1,1′-Biphenyl]-4,4′-diylbis[3,6-bis(1,1-dimethyl ethyl)]-9H-carbazole

99%

Synonym(s):

4,4′-Bis(3,6-di-tert-butyl-9H-carbazol-9-yl)-1,1′-biphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H56N2
CAS Number:
Molecular Weight:
709.01
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

solid

mp

437-442 °C

SMILES string

CC(C)(C)c1ccc2n(-c3ccc(cc3)-c4ccc(cc4)-n5c6ccc(cc6c7cc(ccc57)C(C)(C)C)C(C)(C)C)c8ccc(cc8c2c1)C(C)(C)C

InChI

1S/C52H56N2/c1-49(2,3)35-17-25-45-41(29-35)42-30-36(50(4,5)6)18-26-46(42)53(45)39-21-13-33(14-22-39)34-15-23-40(24-16-34)54-47-27-19-37(51(7,8)9)31-43(47)44-32-38(52(10,11)12)20-28-48(44)54/h13-32H,1-12H3

InChI key

OFUWRAFMFGLINE-UHFFFAOYSA-N

Application

Host materials for efficient solution-processed red and green phosphorescent devices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng-Huan Ho, et al.
Thin Solid Films, 517, 943-947 (2008)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service