Skip to Content
MilliporeSigma
All Photos(1)

Documents

768294

Sigma-Aldrich

Diisooctyl phthalate-d4

98 atom % D, 96% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D4H34O4
CAS Number:
Molecular Weight:
394.58
UNSPSC Code:
85151701

isotopic purity

98 atom % D

Assay

96% (CP)

form

liquid

mass shift

M+4

InChI

1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3/i13D,14D,17D,18D

InChI key

MQIUGAXCHLFZKX-AMEAAFLOSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service