Skip to Content
MilliporeSigma
All Photos(1)

Documents

767867

Sigma-Aldrich

6-Fluoro-3,4-dihydro-2H-quinoline-1-carboxaldehyde

97%

Synonym(s):

6-Fluoro-3,4-dihydro-1(2H)-quinolinecarboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10FNO
CAS Number:
Molecular Weight:
179.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

85-87 °C

SMILES string

Fc1ccc2N(CCCc2c1)C=O

InChI

1S/C10H10FNO/c11-9-3-4-10-8(6-9)2-1-5-12(10)7-13/h3-4,6-7H,1-2,5H2

InChI key

XJTRLWYQQNGUMV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service