Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767557

Sigma-Aldrich

Poly(ethylene glycol) methyl ether azide

average Mn 400

Synonym(s):

Methoxypolyethylene glycol azide, PEG N3, azido mPEG, mPEG azide, mPEG-N3, methoxyPEG azide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

mol wt

PEG average Mn 350 (n~8)
average Mn 400

refractive index

n20/D 1.458

density

1.105 g/mL at 25 °C

Ω-end

azide

α-end

methoxy

storage temp.

2-8°C

Application

PEG functionalized with azide is commonly used in copper-mediated ligation (click chemistry). PEG of this molecular weight should remain amorphous at room temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service