Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

767220

Sigma-Aldrich

Polyester bis-MPA dendron, 2 hydroxyl, 1 azide

generation 1, 97%

Synonym(s):

bisMPA dendrimer, bisMPA dendron

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

hydroxyl surface groups

Assay

97%

form

liquid

mol wt

generation 1

feature

focal point azide

no. Surface Groups

2

refractive index

n20/D 1.478

density

1.120 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CC(CO)(CO)C(=O)OCCCCCCN=[N+]=[N-]

InChI

1S/C11H21N3O4/c1-11(8-15,9-16)10(17)18-7-5-3-2-4-6-13-14-12/h15-16H,2-9H2,1H3

InChI key

VIVYDDCPJPJOCJ-UHFFFAOYSA-N

General description

calculated mol. wt. 259.3

Application

Azide dendrons can be coupled to acetylene functionalized molecules, including acetylene dendrons to form asymmetric dendrimers.

Other Notes

Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service