Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

766887

Sigma-Aldrich

3-Iodo-5-nitrobenzoic acid

97%

Synonym(s):

NSC 40643

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4INO4
CAS Number:
Molecular Weight:
293.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

166-170 °C

SMILES string

OC(=O)c1cc(I)cc(c1)[N+]([O-])=O

InChI

1S/C7H4INO4/c8-5-1-4(7(10)11)2-6(3-5)9(12)13/h1-3H,(H,10,11)

InChI key

GFGURBBHVJTXDU-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service