Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

766615

Sigma-Aldrich

Potassium 2-phenoxyphenyltrifluoroboroate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9BF3KO
CAS Number:
Molecular Weight:
276.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

solid

mp

219-224 °C

SMILES string

[K+].F[B-](F)(F)c1ccccc1Oc2ccccc2

InChI

1S/C12H9BF3O.K/c14-13(15,16)11-8-4-5-9-12(11)17-10-6-2-1-3-7-10;/h1-9H;/q-1;+1

InChI key

SJWZSSLBZXLGEC-UHFFFAOYSA-N

Application

Cross-coupling partner in Palladium-catalyzed carbon-carbon bond formation (Suzuki reaction) and has shown recently to undergo radical cyclization under open-flask conditions

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan W Lockner et al.
Organic letters, 13(20), 5628-5631 (2011-09-20)
Practical radical cyclizations using organoboronic acids and trifluoroborates take place in water, open to air, and in a scalable fashion employing catalytic silver nitrate and stoichiometric potassium persulfate. Both Pschorr-type cyclizations and tandem radical cyclization/trap cascades are described, illustrating the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service