Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76657

Sigma-Aldrich

Pentaerythritol tris[3-(1-aziridinyl)propionate]

technical, ≥90% (CHN)

Synonym(s):

TAZO, Xama 7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H33N3O7
CAS Number:
Molecular Weight:
427.49
Beilstein:
6618745
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (CHN)

autoignition temp.

899 °F

refractive index

n20/D 1.496 (lit.)
n20/D 1.496

density

1.158 g/mL at 25 °C (lit.)

SMILES string

OCC(COC(=O)CCN1CC1)(COC(=O)CCN2CC2)COC(=O)CCN3CC3

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service