Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

765058

Sigma-Aldrich

3,6-Bis(N-carbazolyl)-N-ethylcarbazole

99% (HPLC)

Synonym(s):

9′-Ethyl-9,3′:6′,9′′-ter-9H-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H27N3
CAS Number:
Molecular Weight:
525.64
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (HPLC)

form

powder

mp

308-313 °C

λmax

342 nm in THF

SMILES string

CCn1c2ccc(cc2c3cc(ccc13)-n4c5ccccc5c6ccccc46)-n7c8ccccc8c9ccccc79

InChI

1S/C38H27N3/c1-2-39-33-21-19-25(40-35-15-7-3-11-27(35)28-12-4-8-16-36(28)40)23-31(33)32-24-26(20-22-34(32)39)41-37-17-9-5-13-29(37)30-14-6-10-18-38(30)41/h3-24H,2H2,1H3

InChI key

IGNAMAKPDIBFKL-UHFFFAOYSA-N

Application

Host materials for efficient solution-processed blue phosphorescent devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service