Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

76491

Sigma-Aldrich

Pentacontane

≥95.0% (GC)

Synonym(s):

n-Pentacontane

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$581.00
1 G
$634.00

$581.00


Available to ship onApril 04, 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
$581.00
1 G
$634.00

About This Item

Linear Formula:
CH3(CH2)48CH3
CAS Number:
Molecular Weight:
703.34
Beilstein:
1807879
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$581.00


Available to ship onApril 04, 2025Details


Request a Bulk Order

Assay

≥95.0% (GC)

form

flakes

mp

91-95 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C50H102/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-43-45-47-49-50-48-46-44-42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-50H2,1-2H3

InChI key

PFLUOWJPZLHUEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service