Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76480

Sigma-Aldrich

Pentachlorophenol sodium salt

technical, ≥80% (HPLC/NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl5NaO
CAS Number:
Molecular Weight:
288.32
Beilstein:
3639137
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥80% (HPLC/NT)

impurities

≤15% water

mp

≥300 °C

SMILES string

[Na+].[O-]c1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6HCl5O.Na/c7-1-2(8)4(10)6(12)5(11)3(1)9;/h12H;/q;+1/p-1

InChI key

HCJLVWUMMKIQIM-UHFFFAOYSA-M

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Zhgun et al.
PloS one, 15(4), e0230591-e0230591 (2020-04-03)
In this study, we investigated biodeterioration of materials used in tempera painting by analyzing the structure of the microbiome in ancient tempera paintings exhibited in State Tretyakov Gallery, Moscow, Russia. Samples were obtained from 16th-century paintings, including a grand Russian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service