Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

764108

Sigma-Aldrich

4-Phenylbutyric-d11 acid

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6(D)5(C(D)2)3COOH
Molecular Weight:
175.27
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

mass shift

M+11

SMILES string

OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])=O

InChI

1S/C10H12O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2,(H,11,12)/i1D,2D,3D,4D2,5D,6D,7D2,8D2

InChI key

OBKXEAXTFZPCHS-VRWITDQQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G van Straten et al.
Scientific reports, 7(1), 13112-13112 (2017-10-17)
Urea cycle enzyme deficiency (UCED) patients with hyperammonemia are treated with sodium benzoate (SB) and sodium phenylacetate (SPA) to induce alternative pathways of nitrogen excretion. The suggested guidelines supporting their use in the management of hyperammonemia are primarily based on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service