Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

763810

Sigma-Aldrich

Tributyl(pentafluorophenyl)stannane

90%

Synonym(s):

Tributyl(pentafluorophenyl)tin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H27F5Sn
CAS Number:
Molecular Weight:
457.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

90%

form

liquid

reaction suitability

core: tin
reagent type: catalyst
reaction type: Stille Coupling

impurities

3-4% pentafluorobenzene
3-4% tributyltin chloride

density

1.298 g/mL at 25 °C

SMILES string

FC1=C(F)C(F)=C(F)C(F)=C1[Sn](CCCC)(CCCC)CCCC

InChI

1S/C6F5.3C4H9.Sn/c7-2-1-3(8)5(10)6(11)4(2)9;3*1-3-4-2;/h;3*1,3-4H2,2H3;

InChI key

JHHIELWIJDNKDR-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service