Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762776

Sigma-Aldrich

1,12-Bis((1-methylbenzimidazolium)-3-yl)dodecane dibromide

97%

Synonym(s):

3,3′-(Dodecane-1,12-diyl)bis(1-methyl-1H-benzo[d]imidazol-3-ium) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H40Br2N4
Molecular Weight:
592.45
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

mp

192-197 °C

SMILES string

[Br-].[Br-].Cn1c[n+](CCCCCCCCCCCC[n+]2cn(C)c3ccccc23)c4ccccc14

InChI

1S/C28H40N4.2BrH/c1-29-23-31(27-19-13-11-17-25(27)29)21-15-9-7-5-3-4-6-8-10-16-22-32-24-30(2)26-18-12-14-20-28(26)32;;/h11-14,17-20,23-24H,3-10,15-16,21-22H2,1-2H3;2*1H/q+2;;/p-2

InChI key

JYLHYZQULXGKCH-UHFFFAOYSA-L

Application

Highly efficient catalyst for the benzoin reaction

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken-ichi Iwamoto et al.
Organic & biomolecular chemistry, 6(5), 912-915 (2008-02-23)
Benzoin reactions are catalyzed effectively by a methylene-bridged bis(benzimidazolium) salt to yield alpha-hydroxy ketones, and the reactions proceed in water as the aqueous medium under mild conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service