Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762733

Sigma-Aldrich

Ethyl 3-aminothiophene-2-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO2S
CAS Number:
Molecular Weight:
171.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

40-45 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)c1sccc1N

InChI

1S/C7H9NO2S/c1-2-10-7(9)6-5(8)3-4-11-6/h3-4H,2,8H2,1H3

InChI key

VSRVNHPIXCUMIA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service