Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

762636

Sigma-Aldrich

2-Ethynyl-1,3,5-trimethylbenzene

96%

Synonym(s):

(2,4,6-Trimethylphenyl)acetylene, 2-Ethynylmesitylene, Ethynylmesitylene, Mesitylacetylene, Mesitylethyne

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12
CAS Number:
Molecular Weight:
144.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.547

bp

210-215 °C

density

0.921 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cc1cc(C)c(C#C)c(C)c1

InChI

1S/C11H12/c1-5-11-9(3)6-8(2)7-10(11)4/h1,6-7H,2-4H3

InChI key

VBEPMNWLWNKUGJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.0 °F

Flash Point(C)

83.88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service