Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76256

Sigma-Aldrich

2-(Aminomethyl)-2-methyl-1,3-propanediamine

≥95.0% (GC)

Synonym(s):

1,1,1-Tris(aminomethyl)ethane, Ethylidintris(methylamine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H15N3
CAS Number:
Molecular Weight:
117.19
Beilstein:
1734348
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0% (GC)

SMILES string

CC(CN)(CN)CN

InChI

1S/C5H15N3/c1-5(2-6,3-7)4-8/h2-4,6-8H2,1H3

InChI key

UGDSCHVVUPHIFM-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service