Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

760838

Sigma-Aldrich

Allyl tetraisopropylphosphorodiamidite

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$148.00

$148.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$148.00

About This Item

Linear Formula:
[[(CH3)2CH]2N]2POCH2CH=CH2
CAS Number:
Molecular Weight:
288.41
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

$148.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

form

liquid

refractive index

n20/D 1.466 (lit.)
n20/D 1.467

bp

114-117 °C/0.4 mmHg (lit.)

density

0.903 g/mL at 25 °C (lit.)
0.910 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)N(C(C)C)P(OCC=C)N(C(C)C)C(C)C

InChI

1S/C15H33N2OP/c1-10-11-18-19(16(12(2)3)13(4)5)17(14(6)7)15(8)9/h10,12-15H,1,11H2,2-9H3

InChI key

ZTEGHJIXOZLSOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allyl tetraisopropylphosphorodiamidite may be used in the total synthesis of (−)-tetrahydrolipstatin[1], dolabelide C[2], (-)-salicylihalamides A and B.[3]
Phosphitylation agent used in the preparation of oligonucleotides. Selective deallylation is accomplished by palladium catalysis.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.0 °F

Flash Point(C)

86.67 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of dolabelide C: a phosphate-mediated approach.
Hanson PR, et al.
The Journal of Organic Chemistry, 76(11), 4358-4370 (2011)
Phosphate Tether-Mediated Approach to the Formal Total Synthesis of (?)-Salicylihalamides A and B.
Chegondi R, et al.
The Journal of Organic Chemistry, 76(10), 3909-3916 (2011)
A concise, phosphate-mediated approach to the total synthesis of (?)-tetrahydrolipstatin.
Venukadasula PK, et al.
Organic Letters, 12(7), 1556-1559 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service