Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76080

Sigma-Aldrich

Palladium(II) oxide hydrate

70-80% Pd basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
OPd · xH2O
CAS Number:
Molecular Weight:
122.42 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

70-80% Pd basis

form

powder

composition

H2O, 0.5-1.5 mol/mol

reaction suitability

reagent type: catalyst
core: palladium

SMILES string

O=[Pd].[H]O[H]

InChI

1S/H2O.O.Pd/h1H2;;

InChI key

CJCUJYURPLYQOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrogenation catalyst

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service